Tisk…
Anti-ARHGEF7 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB49018-200EA 0 CZK
BIRBORB49018-200 BIRBORB49018-100
Anti-ARHGEF7 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ARHGEF7 (Rho guanine nucleotide exchange factor (GEF) 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat