Tisk…

Anti-TNFRSF10C Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB49032-100EA 0 CZK
BIRBORB49032-100
Anti-TNFRSF10C Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR