Tisk…
Anti-PDGFB Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB49097-200EA 0 CZK
BIRBORB49097-200 BIRBORB49097-100
Anti-PDGFB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PDGFB (platelet-derived growth factor beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat