Tisk…

Anti-ARHGAP25 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB49126-100EA 0 CZK
BIRBORB49126-100 BIRBORB49126-200
Anti-ARHGAP25 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1500: 12000; Immunochemistry: 150: 1200

Type: Primary
Antigen: ARHGAP25 (Rho GTPase activating protein 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR