Tisk…
Anti-ERG Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB49143-200EA 0 CZK
BIRBORB49143-200 BIRBORB49143-100
Anti-ERG Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat