Tisk…

Anti-NQO1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
BIRBORB66484-100EA 0 CZK
BIRBORB66484-100
Anti-NQO1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Western Blot: 1-1000

Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR