Tisk…

Anti-GAPDH Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB66644-100EA 0 CZK
BIRBORB66644-100
Anti-GAPDH Rabbit polyclonal antibody
Protilátky
Western Blot: 1:5000-1:50000

Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR