Tisk…
Anti-V5 Tag Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB66663-100EA 0 CZK
BIRBORB66663-100 BIRBORB66663-20
Anti-V5 Tag Mouse Monoclonal Antibody
Protilátky
Western Blot: 1:3000-1:10000; Immunofluorescence: 1:200
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Objednat