Tisk…
Anti-TUBB Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB66666-100EA 0 CZK
BIRBORB66666-100 BIRBORB66666-20
Anti-TUBB Mouse Monoclonal Antibody
Protilátky
Western Blot: 1:3000-1:10000; Immunochemistry: 1:200
Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Objednat