Tisk…

Anti-IL20 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB66681-100EA 0 CZK
BIRBORB66681-100
Anti-IL20 Mouse Monoclonal Antibody
Protilátky
ELISA: 1:1000-1:10000

Type: Primary
Antigen: IL20 (interleukin 20)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR