Tisk…

Anti-TET2 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB66693-100EA 0 CZK
BIRBORB66693-100
Anti-TET2 Mouse Monoclonal Antibody
Protilátky
Western Blot: 1:1000-1:5000; Immunoprecipitation: 1:50-1:100

Type: Primary
Antigen: TET2 (tet methylcytosine dioxygenase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM (Mouse)
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR