Tisk…

Anti-GST Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB66695-100EA 0 CZK
BIRBORB66695-100
Anti-GST Mouse Monoclonal Antibody
Protilátky
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR