Tisk…

Anti-SATB1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB66710-100EA 0 CZK
BIRBORB66710-100
Anti-SATB1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: SATB1 (SATB homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR