Tisk…

Anti-SAE1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB66758-100EA 0 CZK
BIRBORB66758-100
Anti-SAE1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: SAE1 (SUMO1 activating enzyme subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR