Tisk…

Anti-CDH1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB66765-100EA 0 CZK
BIRBORB66765-100
Anti-CDH1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:1000

Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR