Tisk…

Anti-MFF Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB102998-100EA 0 CZK
BIRBORB102998-100
Anti-MFF Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: MFF (mitochondrial fission factor)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR