Tisk…

Anti-Cytochrome P450 4A11/22 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB125660-100EA 0 CZK
BIRBORB125660-100 BIRBORB125660-200
Anti-Cytochrome P450 4A11/22 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Cytochrome P450 4A11/22
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR