Tisk…

Anti-NKX6.3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB127108-200EA 0 CZK
BIRBORB127108-200 BIRBORB127108-100
Anti-NKX6.3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NKX6.3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR