Anti-EPN2 Rabbit Polyclonal Antibody
: Biorbyt
Type: Primary
Antigen: EPN2 (epsin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
- Catalog No:
- BIRBORB128034-200
- BIRBORB128034-100
- Antigen symbol:
- EPN2
- EPN2
- Antigen name:
- Epsin 2
- Epsin 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
- : ORB128034-200: BIRBORB128034-200
Specifications
Antigen symbolEPN2Antigen nameEpsin 2Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitIsotypeWestern blotYesPurificationpurified by affinity chromatographyConcentration1 mg/mlStorage bufferPBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.Storage temperatureCoolShipping temperatureIce - : ORB128034-100: BIRBORB128034-100
Specifications
Antigen symbolEPN2Antigen nameEpsin 2Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitIsotypeWestern blotYesPurificationpurified by affinity chromatographyConcentration1 mg/mlStorage bufferPBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.Storage temperatureCoolShipping temperatureIce