Tisk…

Anti-FKBP5 Mouse monoclonal antibody unconjugated

Dodavatel: Biorbyt
BIRBORB67334-25EA 0 CZK
BIRBORB67334-25 BIRBORB67334-100
Anti-FKBP5 Mouse monoclonal antibody unconjugated
Protilátky
Western Blot: 1:2000

Type: Primary
Antigen: FKBP5 (FK506 binding protein 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR