Tisk…

Anti-ATAD3A Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB75627-100EA 0 CZK
BIRBORB75627-100
Anti-ATAD3A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ATAD3A (ATPase family, AAA domain containing 3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR