Tisk…

Anti-POLR2A Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB77807-100EA 0 CZK
BIRBORB77807-100
Anti-POLR2A Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: POLR2A (polymerase (RNA) II (DNA directed) polypeptide A, 220kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR