Tisk…

Anti-USP31 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB89203-50EA 0 CZK
BIRBORB89203-50
Anti-USP31 Goat Polyclonal Antibody
Protilátky
Immunochemistry: 3.75 ?g/ml

Type: Primary
Antigen: USP31 (ubiquitin specific peptidase 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR