Tisk…

Anti-LDHB Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB89635-50EA 0 CZK
BIRBORB89635-50
Anti-LDHB Mouse Monoclonal Antibody
Protilátky
Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: LDHB (lactate dehydrogenase B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR