Tisk…

Anti-NOS2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB3523-200EA 28380 CZK
ABCAAB3523-200
Anti-NOS2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR