Tisk…

Anti-IgG Goat Polyclonal Antibody (Biotin)

Dodavatel: Abcam
ABCAAB5886-500EA 14440 CZK
ABCAAB5886-500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR