Tisk…

Anti-COL5 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB7046-100EA 29230 CZK
ABCAAB7046-100
Anti-COL5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR