Tisk…

Anti-IgG Donkey Polyclonal Antibody (Texas Red®)

Dodavatel: Abcam
ABCAAB7059-500EA 11800 CZK
ABCAAB7059-500
Anti-IgG Donkey Polyclonal Antibody (Texas Red®)
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR