Tisk…

Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Abcam
ABCAAB7069-500EA 11800 CZK
ABCAAB7069-500
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR