Tisk…

Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abcam
ABCAAB7093-500EA 11800 CZK
ABCAAB7093-500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR