Tisk…

Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Abcam
ABCAAB7131-500EA 10510 CZK
ABCAAB7131-500
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR