Tisk…

Anti-IgG Goat Polyclonal Antibody (Biotin)

Dodavatel: Abcam
ABCAAB7145-500EA 11800 CZK
ABCAAB7145-500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG (Golden Syrian Hamster) (golden syrian hamster IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR