Tisk…

Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Abcam
ABCAAB7154-500EA 11800 CZK
ABCAAB7154-500
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR