Tisk…

Anti-ITGB3 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: F11]

ABCAAB7162-100EA 19190 CZK
ABCAAB7162-100
Anti-ITGB3 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: F11]
Type: Primary
Antigen: ITGB3 (integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61))
Clonality: monoclonal
Clone: F11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR