Tisk…

Anti-DMD Mouse Monoclonal Antibody [clone: MANDYS8]

ABCAAB7163-100EA 16480 CZK
ABCAAB7163-100
Anti-DMD Mouse Monoclonal Antibody [clone: MANDYS8]
Type: Primary
Antigen: DMD (dystrophin)
Clonality: monoclonal
Clone: MANDYS8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR