Tisk…

Anti-CD51 + CD61 Mouse Monoclonal Antibody [clone: BV3]

Dodavatel: Abcam
ABCAAB7166-50EA 28680 CZK
ABCAAB7166-50
Anti-CD51 + CD61 Mouse Monoclonal Antibody [clone: BV3]
Protilátky
Type: Primary
Antigen: CD51 + CD61
Clonality: monoclonal
Clone: BV3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR