Tisk…

Anti-ITGB1 Mouse Monoclonal Antibody [clone: BV7]

Dodavatel: Abcam
ABCAAB7168-50EA 28250 CZK
ABCAAB7168-50
Anti-ITGB1 Mouse Monoclonal Antibody [clone: BV7]
Protilátky
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone: BV7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR