Tisk…

Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Abcam
ABCAAB7171-500EA 12660 CZK
ABCAAB7171-500
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR