Tisk…

Anti-ICAM1 Mouse Monoclonal Antibody Biotin [clone: HM1]

ABCAAB7223-25EA 17160 CZK
ABCAAB7223-25
Anti-ICAM1 Mouse Monoclonal Antibody Biotin [clone: HM1]
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: HM1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR