Tisk…

Anti-GFAP Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB7260-50EA 29780 CZK
ABCAAB7260-50
Anti-GFAP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR