Tisk…

Anti-KLK13 Rabbit Polyclonal Antibody

ABCAAB7285-50EA 16080 CZK
ABCAAB7285-50
Anti-KLK13 Rabbit Polyclonal Antibody
Type: Primary
Antigen: KLK13 (kallikrein-related peptidase 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR