Tisk…

Anti-XAB2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB7289-50EA 22340 CZK
ABCAAB7289-50
Anti-XAB2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: XAB2 (XPA binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR