Tisk…

Anti-CR2 Mouse Monoclonal Antibody [clone: 1F8]

Dodavatel: Abcam
ABCAAB7290-6EA 22340 CZK
ABCAAB7290-6
Anti-CR2 Mouse Monoclonal Antibody [clone: 1F8]
Protilátky
Type: Primary
Antigen: CR2 (complement component (3d/Epstein Barr virus) receptor 2)
Clonality: monoclonal
Clone: 1F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR