Tisk…

Anti-IgE Mouse Monoclonal Antibody [clone: 5D4]

Dodavatel: Abcam
ABCAAB7295-500EA 22520 CZK
ABCAAB7295-500
Anti-IgE Mouse Monoclonal Antibody [clone: 5D4]
Protilátky
Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone: 5D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR