Tisk…

Anti-Substance P Rat Monoclonal Antibody [clone: M09205]

Dodavatel: Abcam
ABCAAB7340-100EA 21970 CZK
ABCAAB7340-100
Anti-Substance P Rat Monoclonal Antibody [clone: M09205]
Protilátky
Type: Primary
Antigen: Substance P
Clonality: monoclonal
Clone: M09205
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR