Tisk…

Anti-TNFRSF1B Rat Monoclonal Antibody [clone: HM102]

Dodavatel: Abcam
ABCAAB7369-50EA 26870 CZK
ABCAAB7369-50
Anti-TNFRSF1B Rat Monoclonal Antibody [clone: HM102]
Protilátky
Type: Primary
Antigen: TNFRSF1B (tumor necrosis factor receptor superfamily, member 1B)
Clonality: monoclonal
Clone: HM102
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR