Tisk…

Anti-PECAM1 Rat Monoclonal Antibody [clone: MEC 7.46]

Dodavatel: Abcam
ABCAAB7388-50EA 22710 CZK
ABCAAB7388-50
Anti-PECAM1 Rat Monoclonal Antibody [clone: MEC 7.46]
Protilátky
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEC 7.46
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR