Tisk…

Anti-IgA Mouse Monoclonal Antibody [clone: 5D8]

ABCAAB7401-1EA 0 CZK
ABCAAB7401-1
Anti-IgA Mouse Monoclonal Antibody [clone: 5D8]
Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR