Tisk…

Anti-IgA Mouse Monoclonal Antibody [clone: AA5]

ABCAAB7404-500EA 16280 CZK
ABCAAB7404-500
Anti-IgA Mouse Monoclonal Antibody [clone: AA5]
Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone: AA5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR