Tisk…

Anti-ANPEP Mouse Monoclonal Antibody [clone: WM15]

Dodavatel: Abcam
ABCAAB7417-100EA 23530 CZK
ABCAAB7417-100
Anti-ANPEP Mouse Monoclonal Antibody [clone: WM15]
Protilátky
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: monoclonal
Clone: WM15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR